Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112356.2 | XP_013967831.1 | 486 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |