Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_531808.5 | XP_531808.3 | 4 - 5 | 655 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |