Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630495.2 | XP_005630552.1 | 2 - 3 | 1249 | 96 | |
XM_022404758.1 | XP_022260466.1 | 3 - 4 | 1207 | 96 | ||
XM_022404759.1 | XP_022260467.1 | 3 - 4 | 1174 | 96 | ||
XM_022404760.1 | XP_022260468.1 | 3 - 4 | 1174 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |