Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629023.3 | XP_005629080.1 | 2 - 3 | 322 | 91 | |
XM_005629024.3 | XP_005629081.1 | 2 - 3 | 196 | 91 | ||
XM_014119709.2 | XP_013975184.1 | 1 - 2 | 288 | 91 | ||
XM_022427966.1 | XP_022283674.1 | 1 - 2 | 339 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |