Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048084.1 | NP_001041549.1 | 5 - 6 | 577 | 64 | |
GenBank mRNA | AB185964.1 | - | 5 - 6 | 577 | 64 | |
DQ141222.1 | - | 5 - 6 | 552 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |