Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427724.1 | XP_022283432.1 | 2 - 3 | 201 | 70 | |
XM_532539.4 | XP_532539.1 | 2 - 3 | 656 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |