Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426027.1 | XP_022281735.1 | 5 - 6 | 1258 | 77 | |
XM_022426028.1 | XP_022281736.1 | 5 - 6 | 1192 | 77 | ||
XM_532133.6 | XP_532133.2 | 2 - 3 | 507 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |