Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533729.6 | XP_533729.3 | 227 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |