Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628499.3 | XP_005628556.1 | 9 - 10 | 1293 | 67 | |
XM_022427338.1 | XP_022283046.1 | 9 - 10 | 1267 | 67 | ||
XM_532457.6 | XP_532457.3 | 9 - 10 | 1496 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |