Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424045.1 | XP_022279753.1 | 232 | 75 | ||
XM_531629.5 | XP_531629.2 | 232 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |