Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425120.1 | XP_022280828.1 | 425 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |