Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630532.3 | XP_005630589.1 | 11 - 12 | 1393 | 119 | |
XM_014120331.2 | XP_013975806.1 | 10 - 11 | 1528 | 119 | ||
XM_022404792.1 | XP_022260500.1 | 10 - 11 | 1526 | 119 | ||
XM_022404793.1 | XP_022260501.1 | 9 - 10 | 1283 | 119 | ||
XM_022404794.1 | XP_022260502.1 | 10 - 11 | 1150 | 119 | ||
XM_022404795.1 | XP_022260503.1 | 10 - 11 | 1053 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |