Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638806.4 | XP_003638854.1 | 3 - 4 | 671 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |