Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC23L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619408.3 | XP_005619465.1 | 2 - 3 | 219 | 62 | |
XM_005619409.3 | XP_005619466.1 | 2 - 3 | 322 | 62 | ||
XM_014113150.2 | XP_013968625.2 | 3 - 4 | 361 | 62 | ||
XM_022417938.1 | XP_022273646.1 | 2 - 3 | 293 | 62 | ||
XM_536506.6 | XP_536506.3 | 2 - 3 | 297 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |