Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112008.2 | XP_013967483.1 | 502 | 84 | ||
XM_014112016.2 | XP_013967491.1 | 452 | 84 | |||
XM_022416508.1 | XP_022272216.1 | 616 | 84 | |||
XM_536287.5 | XP_536287.3 | 616 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |