Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639053.4 | XP_003639101.1 | 318 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Inventoried |