Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618279.1 | XP_005618336.1 | 2 - 3 | 228 | 102 | |
XM_022416832.1 | XP_022272540.1 | 2 - 3 | 228 | 102 | ||
XM_022416834.1 | XP_022272542.1 | 2 - 3 | 244 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |