Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536257.6 | XP_536257.1 | 4 - 5 | 2863 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |