Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537955.5 | XP_537955.2 | 2 - 3 | 211 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |