Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624552.2 | XP_005624609.1 | 340 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |