Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620056.2 | XP_005620113.1 | 3 - 4 | 689 | 80 | |
XM_005620057.3 | XP_005620114.1 | 3 - 4 | 726 | 80 | ||
XM_014113605.2 | XP_013969080.1 | 2 - 3 | 784 | 80 | ||
XM_014113606.2 | XP_013969081.1 | 3 - 4 | 689 | 80 | ||
XM_536644.6 | XP_536644.3 | 2 - 3 | 556 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |