Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112800.1 | XP_013968275.1 | 5 - 6 | 620 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |