Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639022.3 | XP_003639070.1 | 10 - 11 | 1721 | 63 | |
XM_022418821.1 | XP_022274529.1 | 9 - 10 | 1488 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |