Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622028.3 | XP_005622085.1 | 6 - 7 | 1301 | 110 | |
XM_537104.6 | XP_537104.3 | 6 - 7 | 1306 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |