Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620674.3 | XP_005620731.1 | 1 - 2 | 136 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |