Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114993.2 | XP_013970468.2 | 261 | 143 | ||
XM_022420878.1 | XP_022276586.1 | 310 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |