Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120828.2 | XP_013976303.1 | 6 - 7 | 969 | 87 | |
XM_022426285.1 | XP_022281993.1 | 6 - 7 | 969 | 87 | ||
XM_541713.6 | XP_541713.3 | 6 - 7 | 1021 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |