Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405523.1 | XP_022261231.1 | 4 - 5 | 543 | 100 | |
XM_022405524.1 | XP_022261232.1 | 4 - 5 | 558 | 100 | ||
XM_540784.7 | XP_540784.3 | 3 - 4 | 287 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |