Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406848.1 | XP_022262556.1 | 4 - 5 | 776 | 61 | |
XM_022406849.1 | XP_022262557.1 | 5 - 6 | 914 | 61 | ||
XM_541935.6 | XP_541935.5 | 5 - 6 | 917 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |