Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426018.1 | XP_022281726.1 | 2 - 3 | 302 | 91 | |
XM_538901.5 | XP_538901.2 | 2 - 3 | 668 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |