Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426018.1 | XP_022281726.1 | 4 - 5 | 632 | 73 | |
XM_538901.5 | XP_538901.2 | 4 - 5 | 998 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |