Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014122213.2 | XP_013977688.1 | 2 - 3 | 401 | 79 | |
XM_022406841.1 | XP_022262549.1 | 2 - 3 | 276 | 79 | ||
XM_022406842.1 | XP_022262550.1 | 2 - 3 | 276 | 79 | ||
XM_022406843.1 | XP_022262551.1 | 2 - 3 | 276 | 79 | ||
XM_022406844.1 | XP_022262552.1 | 2 - 3 | 200 | 79 | ||
XM_022406845.1 | XP_022262553.1 | 2 - 3 | 276 | 79 | ||
XM_541933.6 | XP_541933.3 | 2 - 3 | 276 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |