Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119797.1 | XP_013975272.1 | 636 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |