Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404777.1 | XP_022260485.1 | 3 - 4 | 442 | 85 | |
XM_540212.5 | XP_540212.3 | 3 - 4 | 397 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |