Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629684.3 | XP_005629741.1 | 1932 | 127 | ||
XM_014119874.2 | XP_013975349.1 | 1379 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |