Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAC3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631495.3 | XP_005631552.1 | 2 - 3 | 667 | 79 | |
XM_540873.6 | XP_540873.2 | 1 - 2 | 721 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |