Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627826.2 | XP_005627883.1 | 2038 | 153 | ||
XM_005627827.3 | XP_005627884.1 | 1973 | 153 | |||
XM_022426581.1 | XP_022282289.1 | 2039 | 153 | |||
XM_539103.5 | XP_539103.4 | 2038 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |