Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120645.2 | XP_013976120.1 | 5 - 6 | 753 | 126 | |
XM_541643.6 | XP_541643.2 | 5 - 6 | 681 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |