Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538738.6 | XP_538738.1 | 2 - 3 | 897 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |