Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639501.3 | XP_003639549.1 | 4 - 5 | 799 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |