Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313837.1 | NP_001300766.1 | 11 - 12 | 894 | 83 | |
XM_022421052.1 | XP_022276760.1 | 12 - 13 | 988 | 83 | ||
XM_022421090.1 | XP_022276798.1 | 11 - 12 | 943 | 83 | ||
XM_022421113.1 | XP_022276821.1 | 12 - 13 | 1037 | 83 | ||
XM_022421139.1 | XP_022276847.1 | 8 - 9 | 424 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |