Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_542706.5 | XP_542706.2 | 1 - 2 | 752 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |