Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619711.3 | XP_005619768.1 | 148 | 143 | ||
XM_014113380.2 | XP_013968855.1 | 148 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |