Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128837.1 | NP_001122309.1 | 2 - 3 | 331 | 122 | |
XM_014111141.2 | XP_013966616.1 | 2 - 3 | 537 | 122 | ||
GenBank mRNA | EU373508.1 | - | 2 - 3 | 331 | 122 | |
KT361519.1 | - | 2 - 3 | 331 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |