Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638288.3 | XP_005638345.1 | 5 - 6 | 834 | 79 | |
XM_014109622.2 | XP_013965097.1 | 5 - 6 | 931 | 79 | ||
XM_022412673.1 | XP_022268381.1 | 6 - 7 | 946 | 79 | ||
XM_022412674.1 | XP_022268382.1 | 5 - 6 | 835 | 79 | ||
XM_544638.6 | XP_544638.3 | 5 - 6 | 713 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |