Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544722.6 | XP_544722.2 | 3 - 4 | 422 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |