Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639374.3 | XP_005639431.1 | 766 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |