Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_545546.6 | XP_545546.1 | 2 - 3 | 464 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |