Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544551.5 | XP_544551.4 | 4 - 5 | 825 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |