Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636071.2 | XP_005636128.1 | 1 | 225 | 55 | |
XM_014108026.2 | XP_013963501.1 | 1 | 227 | 55 | ||
XM_543346.5 | XP_543346.2 | 1 | 225 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |